missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD72 Antibody (982405), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5405CL1
This item is not returnable.
View return policy
Description
CD72 Monoclonal antibody specifically detects CD72 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| CD72 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD72, Arg117-Asp359, Accession # P21854 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 971 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 982405 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction