missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD72 Antibody (BU40), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64350MFV450
This item is not returnable.
View return policy
Description
CD72 Monoclonal antibody specifically detects CD72 in Human, Macaque samples. It is validated for Flow Cytometry
Specifications
| CD72 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Macaque | |
| Purified |
| Flow Cytometry | |
| BU40 | |
| Flow Cytometry | |
| CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2 | |
| HF B1 plasmacytoid cell line. Considered to be late B like. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 971 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction