missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD74 Antibody (2D1B3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 217.00 - € 564.00
Specifications
| Antigen | CD74 |
|---|---|
| Clone | 2D1B3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633326
|
Novus Biologicals
NBP2-37341-0.025ml |
0.025 mL |
€ 217.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18116108
|
Novus Biologicals
NBP2-37341 |
0.1 mL |
€ 564.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD74 Monoclonal specifically detects CD74 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
| CD74 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
| CD74 | |
| IgG1 | |
| Affinity Purified |
| 2D1B3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| PBS with 0.05% Sodium Azide | |
| 972 | |
| Purified recombinant fragment of human CD74 (AA: 1-106) expressed in E. Coli. | |
| Primary | |
| 33.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title