missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD74 Antibody (332516) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB35901A
This item is not returnable.
View return policy
Description
CD74 Monoclonal antibody specifically detects CD74 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CD74 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 332516 | |
| Flow Cytometry, CyTOF-ready | |
| CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
| Chinese hamster ovary cell line CHO-S-derived recombinant human CD74, Gln73-Met232, Accession # NP_004346 | |
| 0.1 mL | |
| Immunology | |
| 972 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction