Learn More
CD74 Antibody (332516) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB35901JF669
Product Code. 30176974
  • ONLINE EXCLUSIVE PRICE
    € 628.00
    € 593.25 / 0.10mL
    Save € 34.75
    5% Off
Description
CD74 Monoclonal antibody specifically detects CD74 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD74
Monoclonal
Janelia Fluor 669
CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33
Chinese hamster ovary cell line CHO-S-derived recombinant human CD74, Gln73-Met232, Accession # NP_004346
0.1 mL
Immunology
972
Store at 4°C in the dark.
IgG2b
Flow Cytometry, CyTOF
332516
50mM Sodium Borate
Mouse
Protein A or G purified