missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD74 Antibody (PIN.1), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-1985MFV450
This item is not returnable.
View return policy
Description
CD74 Monoclonal antibody specifically detects CD74 in Human, Mouse (Negative) samples. It is validated for Flow Cytometry
Specifications
| CD74 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative) | |
| Purified |
| Flow Cytometry | |
| PIN.1 | |
| Flow Cytometry | |
| CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
| Peptide corresponding to residues 12-27 of CD74, human invariant chain (short form). | |
| 0.1 mL | |
| Immunology | |
| 972 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction