missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD74 Antibody (SPM523), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34778H
This item is not returnable.
View return policy
Description
CD74 Monoclonal specifically detects CD74 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD74 | |
| Monoclonal | |
| HRP | |
| CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| CD74 is a type II transmembrane protein which binds to the peptide binding groove of newly synthesized MHC class II alpha/beta heterodimers and prevents their premature association with endogenous polypeptides. CD74 is expressed primarily by antigen presenting cells, such as B-lymphocytes (from before the pre-B cell stage to before the plasma cell stage), macrophages, and monocytes, and many epithelial cells. Anti-CD74 stains predominantly germinal center lymphocytes and B-cell lymphomas, but rarely T-cell lymphomas. Anti-CD74 has been shown to be useful in differentiating atypical fibroxanthoma (-) from malignant fibrous histiocytoma (+). | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoprecipitation | |
| SPM523 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CD74 | |
| SU-DHL-4 lymphoma cells | |
| 0.1 mL | |
| Immunology | |
| 972 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction