Learn More
Invitrogen™ CD74 Monoclonal Antibody (PIN.1), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545124
Description
1 μg/mL of MA5-45124 was sufficient for detection of CD74 in 20 μg of PALA cell lysates by colorimetric immunolot analysis using goat anti-mouse IgG: AP as the secondary antibody. Detects approximately 33-35kDa protein doublet corresponding to the molecular mass of the p33 and p35 forms of human CD74.
HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of a 36 kDa alpha chain (DRA) and 27 kDa beta chain (DRB). The alpha chain gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Hundreds of DRB1 alleles have been described and typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. HLA-DR is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides, present them to T-helper cells, and have a critical role in the initiation of the immune response.
Specifications
| CD74 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.2 | |
| P04233, P04441 | |
| CD74 | |
| Human CD74 invariant chain synthetic peptide. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| PIN.1 | |
| APC | |
| CD74 | |
| CD74; CD74 antigen; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 molecule; Cd74 molecule, major histocompatibility complex, class II invariant chain; class II-associated invariant chain peptide; CLIP; DHLAG; dinucleotide microsatellite; gamma chain of class II antigens; H-2 class II histocompatibility antigen gamma chain; histocompatibility: class II antigens, gamma chain of; HLA class II histocompatibility antigen gamma chain; HLADG; HLA-DR antigens-associated invariant chain; HLA-DR-gamma; ia antigen-associated invariant chain; Ia-associated invariant chain; Ia-GAMMA; Ii; invariant polypeptide of major histocompatibility complex, class II antigen-associated; INVG34; MHC class II-associated invariant chain; MHC HLA-DR gamma chain; p33 | |
| Mouse | |
| Protein G | |
| RUO | |
| 16149, 972 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.