missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD77 Synthase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 684.00
Specifications
| Antigen | CD77 Synthase |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18473482
|
Novus Biologicals
NBP1-90824-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18488251
|
Novus Biologicals
NBP1-90824 |
0.1 mL |
€ 684.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD77 Synthase Polyclonal antibody specifically detects CD77 Synthase in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| CD77 Synthase | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9NPC4 | |
| 53947 | |
| IgG | |
| Immunogen affinity purified |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| A14GALTalpha 1,4-galactosyltransferase (globotriaosylceramide synthase, P blood group), A4GALT1, alpha 1,4-galactosyltransferase, Alpha-1,4-galactosyltransferase, Alpha-1,4-N-acetylglucosaminyltransferase, alpha4Gal-T1, CD77 synthase, EC 2.4.1.228, Gb3 synthase, Gb3S, Globotriaosylceramide synthase, lactosylceramide 4-alpha-galactosyltransferase, P blood group (P one antigen), P(k), P(k) antigen synthase, P1, P1/Pk synthase, PK, UDP-galactose:beta-D-galactosyl-beta1-R 4-alpha-D-galactosyltransferase | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:WEPYLLPVLSDASRIALMWKFGGIYLDTDFIVLKNLRNLTNVLGTQSRYVLNGAFLAFERRHEFMALCMRDFVDHYNGWIWGHQGPQLLTRVFKKWCSIRSLAESRACRGVTTLPPEAFYPIPWQDWKKYFEDINPEELPRLLSATYAVH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title