missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79a Mouse, Clone: HM57, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 510.00
Specifications
Antigen | CD79a |
---|---|
Clone | HM57 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941298
|
EnquireBio
973-MSM7-P1 |
100 μg |
€ 510.00
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931298
|
EnquireBio
973-MSM7-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CD79a | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
973 | |
A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein. | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
44kDa |
HM57 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_067612 | |
CD79A | |
IgG1 κ | |
Protein A/G Purified | |
Cell surface | |
Daudi or Ramos cells. Germinal center B-cells in a lymph node or tonsil. |