missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79A Rabbit anti-Human, Clone:RM297, Abnova™
Rabbit monoclonal antibody raised against human CD79A.
Brand: Abnova MAB16923.100uL
This item is not returnable.
View return policy
Description
The B lymphocyte antigen receptor is a multimeric complex that includes the antigen-specific component, surface immunoglobulin (Ig). Surface Ig non-covalently associates with two other proteins, Ig-alpha and Ig-beta, which are necessary for expression and function of the B-cell antigen receptor. This gene encodes the Ig-alpha protein of the B-cell antigen component. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]Specifications
| CD79A | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-200) Western Blot (1:100-200) The optimal working dilution should be determined by the end user. |
|
| CD79A | |
| Rabbit | |
| 100 μL | |
| This antibody reacts to human CD79a. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| RM297 | |
| CD79a molecule, immunoglobulin-associated alpha | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| IGA, MB-1 | |
| A peptide corresponding to C-terminus of human CD79A. | |
| 973 | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction