Learn More
CD79B Antibody (003), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90705MFV500
Product Code. 18478439
  • € 629.00 / 0.10mL
Description
CD79B Monoclonal antibody specifically detects CD79B in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry (Paraffin)
50mM Sodium Borate
974
Purified
003
Primary
Store at 4C in the dark.