missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79B Antibody (CB3-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28528
This item is not returnable.
View return policy
Description
CD79B Monoclonal specifically detects CD79B in Human samples. It is validated for Flow Cytometry.
Specifications
| CD79B | |
| Monoclonal | |
| 0.1 mg/mL | |
| Flow Cytometry <= 1 ug/10^6 cells | |
| CD79B | |
| Ramos B cells | |
| 0.1 mg | |
| Immunology | |
| 974 | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| CB3-1 | |
| Unconjugated | |
| B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human CD79 beta/B29/Ig beta | |
| Store at 4C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction