missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79B Antibody (HM79-11), DyLight 680, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NB100-65611FR
This item is not returnable.
View return policy
Description
CD79B Monoclonal specifically detects CD79B in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD79B | |
| Monoclonal | |
| DyLight 680 | |
| B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
| Armenian Hamster | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-65611 recognizes murine CD79 beta, which is expressed by B cells as part of the B cell receptor complex (immunoglobulin and the CD79 alpha/beta heterodimer). Murine CD79 beta is expressed at the cell surface of pro-B cells prior to surface immunoglobulin, and is expressed throughout B cell differentiation. CD79 beta is a B cell specific marker, valuable for the detection of B cells at all maturation stages. | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| HM79-11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD79B | |
| CD79 alpha/CD79 beta heterodimer purified from WEH1231 B cells. | |
| 0.1 mL | |
| Immunology | |
| 974 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction