missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79B Antibody (IGB/1844), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08597B
This item is not returnable.
View return policy
Description
CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for ELISA, Protein Array
Specifications
| CD79B | |
| Monoclonal | |
| Biotin | |
| B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
| Recombinant human CD79B protein fragment (around aa 29-159) (exact sequence is proprietary) (Uniprot: P40259) | |
| 100 μg | |
| Immunology | |
| 974 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Protein Array | |
| IGB/1844 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction