missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79b (B-Cell Marker) Mouse, Clone: IGB/1844, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 478.00
Specifications
| Antigen | CD79b (B-Cell Marker) |
|---|---|
| Clone | IGB/1844 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15991298
|
EnquireBio
974-MSM4-P0 |
20 μg | |||||||
|
15901308
|
EnquireBio
974-MSM4-P1 |
100 μg |
€ 554.40 € 478.00 / 100µg Save € 76.40 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD79b (B-Cell Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human CD79b protein fragment (apprx. aa29-159) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 39kDa |
| IGB/1844 | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| RUO | |
| 974 | |
| IgG1 κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Cell surface | |
| Daudi or Ramos cells. Germinal center B-cells in a lymph node or tonsil. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title