missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79B Monoclonal Antibody (OTI2B6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810463S
This item is not returnable.
View return policy
Description
CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CD79B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P40259 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 974 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B6 | |
| Unconjugated | |
| Cd79b | |
| AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7 | |
| Full length human recombinant protein of human CD79B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction