missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79B Mouse anti-Human, Clone: OTI4E11, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810480
This item is not returnable.
View return policy
Description
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.Specifications
| CD79B | |
| Monoclonal | |
| Unconjugated | |
| CD79B | |
| AGM6, B29, IGB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 974 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4E11 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P40259 | |
| CD79B | |
| Full length human recombit protein of human CD79B produced in HEK293T. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction