Learn More
CD8 Antibody (12.C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NB100-64021
 View more versions of this product
Product Code. 18734963
  • € 526.00 / 0.10mg
Description
CD8 Monoclonal specifically detects CD8 in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD8
Monoclonal
1.0 mg/mL
Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
CD8A
Made to Rabbit CD8
0.1 mg
Innate Immunity, Stem Cell Markers
925
Rabbit
Purified
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
12.C7
Unconjugated
CD8, CD8a molecule, LEU2, p32
Mouse
Protein G purified
RUO
Primary
Recognizes the rabbit CD8 cell surface antigen, expressed by a subset of T lymphocytes with cytotoxic/suppressor activity.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.