missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (14) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.00 - € 351.00
Specifications
| Antigen | CD8 |
|---|---|
| Clone | 14 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607756
|
Novus Biologicals
NBP2-50467-0.025ml |
0.025 mL |
€ 201.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669638
|
Novus Biologicals
NBP2-50467 |
0.1 mL |
€ 351.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOFSpecifications
| CD8 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 925 | |
| IgG1 | |
| Protein A purified |
| 14 | |
| Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2 | |
| CD8 Antibody (14) was developed against Fibronectin-purified human monocytes | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title