missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (fCD8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28254AF532
This item is not returnable.
View return policy
Description
CD8 Monoclonal specifically detects CD8 in Feline samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
| CD8 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD8A | |
| Feline CD8, Mr 70 kDa (38/31 kDa under reducing conditions). | |
| Primary | |
| Feline CD8, Mr 70 kDa (38/31 kDa under reducing conditions). | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry | |
| fCD8 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry | |
| CD8, CD8a molecule, LEU2, p32 | |
| Mouse | |
| 0.1 mL | |
| 925 | |
| Feline | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction