Learn More
CD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand:  Novus Biologicals NBP2-29475
 View more versions of this product
Product Code. 18749133
  • € 575.00 / 0.10mg
Description
CD8 Polyclonal specifically detects CD8 in Human, Mouse, Porcine samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD8
CD8, CD8a molecule, LEU2, p32
32 kDa
0.1 mg
Flow Cytometry, Immunofluorescence, Immunoprecipitation
Recombinant fragment from human CD8 C-terminal cytoplasmic domain of alpha chain (exact sequence is proprietary). (Uniprot: P01732)
Protein A purified
Pig