missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (OX-8), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12523APCCY7
This item is not returnable.
View return policy
Description
CD8 Monoclonal specifically detects CD8 in Rat samples. It is validated for Flow Cytometry.
Specifications
| CD8 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CD8A | |
| High Mw glycoproteins from rat thymocytes | |
| 0.1 mL | |
| Primary | |
| The mouse monoclonal antibody OX-8 recognizes the hinge-like membrane-proximal domain of rat CD8a (32-34 kDa; alpha chain of the CD8 antigen). | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OX-8 | |
| Flow Cytometry | |
| CD8, CD8a molecule, LEU2, p32 | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 925.0 | |
| Rat | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction