missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (RPA-T8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25195F
This item is not returnable.
View return policy
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Cell Surface).
Specifications
| CD8 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CD8A | |
| CD8 (CD8a; alpha chain) | |
| 0.1 mL | |
| Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 925 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| RPA-T8 | |
| Flow Cytometry, Immunohistochemistry-Frozen, Flow (Cell Surface) | |
| CD8, CD8a molecule, LEU2, p32 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction