Learn More
CD8 Antibody (V44P1H9*F11), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50152B
Product Code. 18116048
  • ONLINE EXCLUSIVE PRICE
    € 466.00
    € 439.95 / 0.10mL
    Save € 26.05
    5% Off
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
CD8 Monoclonal specifically detects CD8 in Fish samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
CD8
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
CD8A
Ovalbumin-conjugated synthetic peptide
0.1 mL
Innate Immunity
925
Store at 4C in the dark.
IgG1 κ
ELISA
V44P1H9*F11
ELISA
CD8, CD8a molecule, LEU2, p32
Mouse
Protein A purified
RUO
Primary
Fish