missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (YTC182.20), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64922PCP
This item is not returnable.
View return policy
Description
CD8 Monoclonal specifically detects CD8 in Human, Primate samples. It is validated for Flow Cytometry.
Specifications
| CD8 | |
| Monoclonal | |
| PerCP | |
| P01732 | |
| CD8A | |
| Mouse L cells transfected with human CD8 gene | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes the human CD8 cell surface antigen, a 30/32kD glycoprotein expressed by the cytotoxic/suppressor subset of T lymphocytes, and more weakly by NK cells. | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| YTC182.20 | |
| Flow Cytometry | |
| CD8, CD8a molecule, LEU2, p32 | |
| Rat | |
| 26 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 925 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction