missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 beta Antibody (H35-17.2), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43449FR
This item is not returnable.
View return policy
Description
CD8 beta Monoclonal specifically detects CD8 beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD8 beta | |
| Monoclonal | |
| DyLight 680 | |
| CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| H35-17.2 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD8B | |
| The immunogen for this antibody was CD8. | |
| 0.1 mL | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
| 926 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction