missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 beta Antibody (H35-17.2), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43449MFV610
This item is not returnable.
View return policy
Description
CD8 beta Monoclonal antibody specifically detects CD8 beta in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD8 beta | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| H35-17.2 | |
| Flow Cytometry | |
| CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain | |
| The immunogen for this antibody was CD8. | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
| 926 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction