Learn More
CD8 Mouse anti-Human, FITC, Clone: SK1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech FITC-65146-100TESTS
This item is not returnable.
View return policy
Description
The CD8 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8. This antibody recognizes human antigen. The CD8 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
| CD8 | |
| Monoclonal | |
| 5 μL (0.125 μg)/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| SK1 | |
| FITC | |
| 100 Tests | |
| 925 | |
| IgG1 |