missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80 Armenian Hamster, APC, Clone: 16-10A1, enQuireBio™
Armenian Hamster Monoclonal Antibody
€ 156.45 - € 367.50
Specifications
| Antigen | CD80 |
|---|---|
| Clone | 16-10A1 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15922148
|
EnquireBio
QAB50-APC-25ug |
25 μg |
€ 156.45
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15912148
|
EnquireBio
QAB50-APC-100ug |
100 μg |
€ 367.50
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD80 | |
| 0.5mg/mL | |
| Monoclonal | |
| Armenian Hamster | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 12519 | |
| IgG | |
| Protein A/G Purified |
| 16-10A1 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| XP_017172347 | |
| Cd80 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title