missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD80 (B7-1) Monoclonal Antibody (16-10A1), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
€ 218.00 - € 408.00
Specifications
| Antigen | CD80 (B7-1) |
|---|---|
| Clone | 16-10A1 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17885317
|
Invitrogen™
367-0801-80 |
25 μg |
€ 218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17815297
![]() |
Invitrogen™
367-0801-82 |
100 μg |
€ 408.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28, and of an inhibitory receptor CTLA-4 (CD152). Both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, and both molecules can also substitute for each other in this process. CD80 is rapidly induced on the surface of in vitro activated B cells, with Epstein Barr Virus (EBV) transformed B cell lines, with Burkitts lymphoma cell lines, with freshly isolated follicular B lymphoma cells, T cells, and monocytes. The B-lymphocyte activation antigen B7-1 provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 and CTLA4 ligands of T cells. Diseases associated with CD80 dysfunction include gallbladder squamous cell carcinoma and myocarditis.Specifications
| CD80 (B7-1) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Activation B7-1 antigen; B7; B7 protein; B7.1; B71; B7-1; B7-1 protein; B7-1 protein precursor; BB1; B-lymphocyte activation antigen B7; Cd28l; CD28LG; CD28LG1; CD80; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 anitgen; CD80 antigen; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); Cd80 molecule; CD80 protein precursor; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; LAB7; Ly53; Ly-53; MIC17; sCD 80; sCD80; secreted B7-1 protein precursor; soluble CD 80; soluble CD80; T-cell costimulatory molecule; T-cell co-stimulatory protein B7-1; T-lymphocyte activation antigen CD80; TS/A-1; TSA1 | |
| CD80 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD80 |
| 16-10A1 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 737 | |
| Armenian Hamster | |
| Canine, Mouse, Pig | |
| Q00609 | |
| 12519, 397161, 403765 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts