Learn More
CD80 Monoclonal Antibody (OTI1G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA812243S
Encompass_Preferred
 View more versions of this product
Product Code. 17556634
  • € 242.00 / 30µL
Description
CD80 Monoclonal antibody specifically detects CD80 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
CD80
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P33681
Mouse
Affinity chromatography
RUO
941
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Flow Cytometry, Western Blot
OTI1G9
Unconjugated
CD80
Activation B7-1 antigen; B7; B7 protein; B7.1; B71; B7-1; B7-1 protein; B7-1 protein precursor; BB1; B-lymphocyte activation antigen B7; Cd28l; CD28LG; CD28LG1; CD80; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 anitgen; CD80 antigen; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); Cd80 molecule; CD80 protein precursor; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; LAB7; Ly53; Ly-53; MIC17; sCD 80; sCD80; secreted B7-1 protein precursor; soluble CD 80; soluble CD80; T-cell costimulatory molecule; T-cell co-stimulatory protein B7-1; T-lymphocyte activation antigen CD80; TS/A-1; TSA1
Full length human recombinant protein of human CD80 produced in HEK293T cell.
30 μL