missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 Antibody (1D6), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81096CL1
This item is not returnable.
View return policy
Description
CD81 Monoclonal antibody specifically detects CD81 in Chimpanzee,Goat,Human,Sheep samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Functional Assay,Immunocytochemistry/Immunofluorescence
Specifications
| CD81 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| This antibody was produced by immunizing mice with 5A6-aggregated OCI-LY8 B cells. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 975 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 1D6 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Chimpanzee, Goat, Human, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction