missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 Antibody (1D6), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65805PECY55
This item is not returnable.
View return policy
Description
CD81 Monoclonal specifically detects CD81 in Human, Mouse, Goat, Primate, Sheep samples. It is validated for Flow Cytometry.
Specifications
| CD81 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| CD81 | |
| Aggregated OCI-Ly8 human B cell line | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Goat, Primate, Sheep | |
| IgG1 κ |
| Flow Cytometry | |
| 1D6 | |
| Flow Cytometry | |
| CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| Mouse | |
| Protein G purified | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 975.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction