missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 Antibody (431301) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4865JF525
This item is not returnable.
View return policy
Description
CD81 Monoclonal antibody specifically detects CD81 in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| CD81 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| NS0 mouse myeloma cell line transfected with mouse CD81, Met1-Tyr236, Accession # NP_598416 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 431301 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 975 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction