missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 Antibody (M38), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-44859
This item is not returnable.
View return policy
Description
CD81 Monoclonal specifically detects CD81 in Human, Mouse, Feline, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| CD81 | |
| Monoclonal | |
| FITC | |
| P60033 | |
| CD81 | |
| MOLT-4, human T-ALL cell line (NM_004356.3). | |
| Size Exclusion Chromatography | |
| RUO | |
| Primary | |
| The antibody M38 reacts with CD81, a 25 kDa member of the tetraspanin family, expressed on majority of cells. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| M38 | |
| Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Knockout Validated | |
| CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| Mouse | |
| 25 kDa | |
| 100 Tests | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 975 | |
| Human, Feline, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction