Learn More
Invitrogen™ CD81 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585150
Description
Keep as concentrated solution. Predicted reactivity: Mouse (83%), Rat (86%), Pig (92%), Sheep (92%), Rhesus Monkey (96%), Chimpanzee (100%), Bovine (89%). Positive Control: human CD81-transfected 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD81 (TAPA-1, target of anti-proliferative antibody-1) is a member of the tetraspanin family, is expressed on virtually all nucleated cells, but above all on germinal center B cells. CD81 forms complexes with other tetraspanin proteins, integrins, coreceptors, MHC class I and II molecules, and influences adhesion, morphology, activation, proliferation and differentiation of B, T cells. In muscles, CD81 promotes cell fusion and myotube maintenance. CD81 has been also identified as a receptor for the hepatitis C virus. Like members of the tetraspanin family that include CD9, CD37, CD53, CD63, and CD82, CD81 is a cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. CD81 is a cell surface glycoprotein that is known to complex with integrins. CD81 appears to promote muscle cell fusion and support myotube maintenance. CD81 associates with CD19, CD21, Leu 13, and integrins on cell membrane and acts as a receptor for the envelope protein E2 of chronic hepatitis C virus. Antibodies to CD81 have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. CD81 is also localized in the tumor-suppressor gene region and is a candidate gene for malignancies.
Specifications
| CD81 | |
| Polyclonal | |
| Unconjugated | |
| CD81 | |
| 26 kDa cell surface protein TAPA-1; CD 81 antigen; Cd81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 molecule; CVID6; S5.7; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; Tetraspanin28; tetraspanin-28; Tspan28; Tspan-28 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12520, 975 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Neutralization, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P35762, P60033 | |
| CD81 | |
| Recombinant protein encompassing a sequence within the center region of human CD81. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.