missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD82/Kai-1 Antibody (SR1500), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21818-100ul
This item is not returnable.
View return policy
Description
CD82/Kai-1 Monoclonal antibody specifically detects CD82/Kai-1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| CD82/Kai-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| SR1500 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| CD82 antigenC33, CD82 molecule, IA4C33 antigen, Inducible membrane protein R2, KAI1GR15, kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyteantigen, antigen detected by monoclonal and IA4)), Metastasis suppressor Kangai-1, R2, SAR2tetraspanin-27, ST6tspan-27, Suppressor of tumorigenicity 6 protein, Tetraspanin-27, Tspan-27, TSPAN274F9 | |
| A synthesized peptide derived from human CD82/Kai-1 (Uniprot #: P27701) | |
| 100 μg | |
| Cancer | |
| 3732 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction