missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD82/Kai-1 Antibody (TS82b), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18118F
This item is not returnable.
View return policy
Description
CD82/Kai-1 Monoclonal antibody specifically detects CD82/Kai-1 in Human samples. It is validated for Flow Cytometry
Specifications
| CD82/Kai-1 | |
| Monoclonal | |
| FITC | |
| CD82 antigenC33, CD82 molecule, IA4C33 antigen, Inducible membrane protein R2, KAI1GR15, kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyteantigen, antigen detected by monoclonal and IA4)), Metastasis suppressor Kangai-1, R2, SAR2tetraspanin-27, ST6tspan-27, Suppressor of tumorigenicity 6 protein, Tetraspanin-27, Tspan-27, TSPAN274F9 | |
| Jurkat and HEL cell lines. Hybridoma: Myeloma X63/AG8. x Balb/c spleen cells | |
| 0.1 mL | |
| Cancer | |
| 3732 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| TS82b | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction