missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD82 Recombinant Rabbit Monoclonal Antibody (23GB5795), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 217.00 - € 590.00
Specifications
| Antigen | CD82 |
|---|---|
| Clone | 23GB5795 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088768
![]() |
Invitrogen™
MA553926 |
100 μL |
€ 590.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088118
|
Invitrogen™
MA553927 |
20 μL |
€ 217.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD82 Recombinant Monoclonal antibody specifically detects CD82 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CD82 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P27701 | |
| 3732 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5795 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4F9; AA682076; AL023070; C33; C33 antigen; Cd82; CD82 antigen; CD82 antigen (R2 leukocyte antigen antigen detected by monoclonal and antibody IA4)); CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); CD82 molecule; GR15; IA4; Inducible membrane protein R2; KAI1; Kangai 1 (suppression of tumerigenicity 6 prostate) CD82 antigen; kangai 1 (suppression of tumorigenicity 6), prostate; kangai 1 (suppression of tumorigenicity 6, prostate); Kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); metastasis suppressor Kangai-1; Metastasis suppressor Kangai-1 homolog; R2; SAR2; ST6; Suppressor of tumorigenicity 6 protein; Tetraspanin-27; Tspan27; tspan-27 | |
| A synthesized peptide derived from human CD82 (200-267AA). | |
| Primary | |
| -20°C | |
| CD82 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title