missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD83 Antibody (HB15e) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1774JF635
This item is not returnable.
View return policy
Description
CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| CD83 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cells transfected with human CD83 | |
| 0.5 mL | |
| Dendritic Cell Markers | |
| 9308 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| HB15e | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction