missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD84/SLAMF5 Antibody (CD84.1.21), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65929B
This item is not returnable.
View return policy
Description
CD84/SLAMF5 Monoclonal antibody specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry
Specifications
| CD84/SLAMF5 | |
| Monoclonal | |
| Biotin | |
| CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
| Mouse pre-B cell line 300.19 transfected with CD84 cDNA. | |
| 0.1 mL | |
| CD Markers, Immunology, Stem Cells | |
| 8832 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| CD84.1.21 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction