missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD84/SLAMF5 Antibody (SR2045), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 449.00
Specifications
| Antigen | CD84/SLAMF5 |
|---|---|
| Clone | SR2045 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620245
|
Novus Biologicals
NBP3-22473-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644504
|
Novus Biologicals
NBP3-22473-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD84/SLAMF5 Monoclonal antibody specifically detects CD84/SLAMF5 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| CD84/SLAMF5 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
| A synthesized peptide derived from human CD84/SLAMF5 (Uniprot #: Q9UIB8) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2045 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| CD Markers, Immunology, Stem Cells | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 8832 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title