missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD86/CD209/CD83 Mouse anti-Human, APC,PE,PerCP-Cy™,¢5.5, Clone: HB15e,2331 (FUN-1),DCN46, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 334098
This item is not returnable.
View return policy
Description
CD86/CD209/CD83 Monoclonal antibody specifically detects CD86/CD209/CD83 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD86/CD209/CD83 | |
| Monoclonal | |
| APC/PE/PerCP-Cyanine5.5 | |
| 50 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ, IgG2b κ |
| Flow Cytometry | |
| HB15e, 2331 (FUN-1), DCN46 | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction