Learn More
CD86 Mouse, anti-Rat, CoraLite Plus 750, Clone: 24F, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL750-65224-100UG
Description
The CD86 antibody from Proteintech is a mouse monoclonal antibody generated with htlv-1-transformed lewis-s1 rat t cell line of rat CD86. This antibody recognizes rat antigen. The CD86 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 750.
Specifications
| CD86 | |
| 24F | |
| CoraLite Plus 750 | |
| Mouse | |
| Primary | |
| Rat | |
| 155 kDa | |
| IgG1 κ |
| Flow Cytometry | |
| 500 μg/mL | |
| B7 2, Cd86, CD86 molecule | |
| 100 μg | |
| 56822 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.