Learn More
CD86 Rat, APC, Clone: GL-1 (GL1), enQuireBio™
Rat Monoclonal Antibody
€ 156.45 - € 367.50
Specifications
| Antigen | CD86 |
|---|---|
| Clone | GL-1 (GL1) |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15912158
|
EnquireBio
QAB51-APC-25ug |
25 μg |
€ 156.45
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15902158
|
EnquireBio
QAB51-APC-100ug |
100 μg |
€ 367.50
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD86 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 12524 | |
| IgG2a κ | |
| Protein A/G Purified |
| GL-1 (GL1) | |
| Flow Cytometry | |
| APC | |
| RUO | |
| NP_062261 | |
| Cd86 | |
| Primary | |
| Store at 2-4°C. |