missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD86 Recombinant Rabbit Monoclonal Antibody (313), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 109.00 - € 326.00
Specifications
| Antigen | CD86 |
|---|---|
| Clone | 313 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019704
![]() |
Invitrogen™
MA552361 |
100 Tests |
€ 326.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019724
|
Invitrogen™
MA552362 |
25 Tests |
€ 109.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD86 Recombinant Monoclonal antibody specifically detects CD86 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD86 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.03% ProClin 300 | |
| activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2 | |
| Recombinant Mouse CD86 Protein (Val 24-Glu245). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD86 |
| 313 | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Mouse | |
| P42082 | |
| 12524 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title