missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8A Mouse anti-Human, Clone: 143-44 (DY-634), Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13763.100Reactions
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against native human CD8A.
- High specificity
- High reactivity
Specifications
| CD8A | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| P01732 | |
| CD8A | |
| T-lymphocyte differentiation antigen. | |
| 100 Reactions | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 143-44 | |
| Mouse monoclonal antibody raised against native human CD8A. | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| CD8/Leu2/MAL/p32 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 925 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only (RUO)
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu