Learn More
CD8a Mouse anti-Human, CoraLite 647, Clone: RPA-T8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL647-65144-100TESTS
This item is not returnable.
View return policy
Description
The CD8a antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8a. This antibody recognizes human antigen. The CD8a antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Specifications
| CD8a | |
| Monoclonal | |
| 5 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| RPA-T8 | |
| CoraLite 647 | |
| 100 Tests | |
| 925 | |
| IgG1 |