Learn More
CD8a Mouse anti-Human, CoraLite 647, Clone: RPA-T8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL647-65144-100TESTS
Product Code. 16891146
  • € 144.00 / 100 tests
Description
The CD8a antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8a. This antibody recognizes human antigen. The CD8a antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Specifications
Specifications
CD8a
Monoclonal
5 μL/Test
Mouse
Primary
Human
Flow Cytometry
RPA-T8
CoraLite 647
100 Tests
925