Learn More
CD8a Mouse, FITC, Clone: Hit8a, enQuireBio™
Mouse Monoclonal Antibody
€ 116.55 - € 156.45
Specifications
| Antigen | CD8a |
|---|---|
| Clone | Hit8a |
| Concentration | 5μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15921658
|
EnquireBio
QAB20-F-25Tests |
25 Tests |
€ 116.55
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15911658
|
EnquireBio
QAB20-F-100Tests |
100 Tests |
€ 156.45
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD8a | |
| 5μL/Test | |
| Monoclonal | |
| Mouse | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 925 | |
| IgG1 κ | |
| Protein A/G Purified |
| Hit8a | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| NP_741969 | |
| CD8A | |
| Primary | |
| Store at 2-4°C. |